Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTAATAAAGACAAAAAAAGTGT[C/T]CTACAGAGGAGCAATTATGGTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMPD3 - FERM and PDZ domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032428.1 | Intron | NP_115804.1 | ||||
XM_006724709.2 | Intron | XP_006724772.1 | ||||
XM_011531056.2 | Intron | XP_011529358.1 | ||||
XM_011531057.2 | Intron | XP_011529359.1 | ||||
XM_011531058.2 | Intron | XP_011529360.1 | ||||
XM_017029901.1 | Intron | XP_016885390.1 | ||||
XM_017029902.1 | Intron | XP_016885391.1 |
FRMPD3-AS1 - FRMPD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |