Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTATTTTCTGTATGACCGTGAC[C/T]GCTTTATTTGAAGAGGGAGTGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRK - Nik related kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198465.3 | Intron | NP_940867.2 | ||||
XM_006724632.3 | Intron | XP_006724695.1 | ||||
XM_006724633.3 | Intron | XP_006724696.1 | ||||
XM_006724634.3 | Intron | XP_006724697.1 | ||||
XM_011530887.2 | Intron | XP_011529189.1 |