Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCCCAAGATGGCTGCTGAGGA[C/T]GAGTTACAGCTGCCGCGGCTCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGBP1 - immunoglobulin (CD79A) binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001551.2 | 313 | Silent Mutation | GAC,GAT | D,D 5 | NP_001542.1 | |
XM_017029489.1 | 313 | Silent Mutation | GAC,GAT | D,D 5 | XP_016884978.1 | |
XM_017029490.1 | 313 | Silent Mutation | GAC,GAT | D,D 5 | XP_016884979.1 |