Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTCTTTTAACAGCCTTCTGGCT[G/T]TGAGCCAACAGGAAGAACTAGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300508 | ||||||||||||||||||||
Literature Links: |
UTP14A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UTP14A - UTP14A small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166221.1 | 127 | Missense Mutation | GTG,TTG | V,L 13 | NP_001159693.1 | |
NM_006649.3 | 127 | Missense Mutation | GTG,TTG | V,L 13 | NP_006640.2 | |
XM_017029237.1 | 127 | UTR 5 | XP_016884726.1 | |||
XM_017029238.1 | 127 | UTR 5 | XP_016884727.1 |