Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATTTTTATCCTCTTTTCCATC[G/T]TCTTCCCCTTTTTCATCTTTGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300385 | ||||||||||||||||||||
Literature Links: |
HMGN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGN5 - high mobility group nucleosome binding domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030763.2 | 624 | Missense Mutation | GAA,GAC | E,D 155 | NP_110390.1 | |
XM_006724688.3 | 624 | Missense Mutation | GAA,GAC | E,D 156 | XP_006724751.1 | |
XM_017029839.1 | 624 | Missense Mutation | GAA,GAC | E,D 146 | XP_016885328.1 | |
XM_017029840.1 | 624 | Missense Mutation | GAA,GAC | E,D 145 | XP_016885329.1 |