Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACAGTCATCCCTGAAACCAGC[A/G]AGGAAAATAATGAGGACATCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIH1D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIH1D3 - PIH1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169154.1 | 306 | Missense Mutation | AAG,GAG | K,E 82 | NP_001162625.1 | |
NM_173494.1 | 306 | Missense Mutation | AAG,GAG | K,E 82 | NP_775765.1 | |
XM_011530855.2 | 306 | Missense Mutation | AAG,GAG | K,E 82 | XP_011529157.1 |