Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGGAAAAGATCTCCACCTTGG[C/G]AATCATTATACATATTGTGATCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300823 | ||||||||||||||||||||
Literature Links: |
IDS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDS - iduronate 2-sulfatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000202.7 | 1789 | Missense Mutation | TCC,TGC | S,C 539 | NP_000193.1 | |
NM_001166550.3 | 1789 | Missense Mutation | TCC,TGC | S,C 449 | NP_001160022.1 | |
NM_006123.4 | 1789 | Intron | NP_006114.1 |