Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTTTGTTGTTGCTGTTGAGAC[G/T]TAGGAGTTTTTTAATATGTTCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300171 | ||||||||||||||||||||
Literature Links: |
MTMR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTMR1 - myotubularin related protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306144.1 | Intron | NP_001293073.1 | ||||
NM_001306145.1 | Intron | NP_001293074.1 | ||||
NM_003828.3 | Intron | NP_003819.1 | ||||
XM_005274765.3 | Intron | XP_005274822.1 | ||||
XM_005274766.3 | Intron | XP_005274823.1 | ||||
XM_005274767.3 | Intron | XP_005274824.1 | ||||
XM_006724855.3 | Intron | XP_006724918.1 | ||||
XM_011531209.2 | Intron | XP_011529511.1 | ||||
XM_011531210.2 | Intron | XP_011529512.1 | ||||
XM_017029920.1 | Intron | XP_016885409.1 | ||||
XM_017029921.1 | Intron | XP_016885410.1 | ||||
XM_017029922.1 | Intron | XP_016885411.1 | ||||
XM_017029923.1 | Intron | XP_016885412.1 | ||||
XM_017029924.1 | Intron | XP_016885413.1 |