Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGATGATTCGGAACTGGAACTG[C/G]AGGGCGAGCTGGAGCAAGATTCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300112 MIM: 300111 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGIX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGIX - MAGI family member, X-linked | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLP2 - proteolipid protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002668.2 | 1752 | Intron | NP_002659.1 |
PRICKLE3 - prickle planar cell polarity protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307979.1 | 1752 | Missense Mutation | TCC,TGC | S,C 482 | NP_001294908.1 | |
NM_006150.4 | 1752 | Missense Mutation | TCC,TGC | S,C 550 | NP_006141.2 |