Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACGCGGGTGGACAAGTGTACC[C/T]GGGAGAGGCATCCCGGGTATCGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300927 | ||||||||||||||||||||
Literature Links: |
PRDX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRDX4 - peroxiredoxin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006406.1 | 225 | Missense Mutation | CCG,CTG | P,L 61 | NP_006397.1 | |
XM_005274438.1 | 225 | Intron | XP_005274495.1 | |||
XM_017029230.1 | 225 | Missense Mutation | CCG,CTG | P,L 61 | XP_016884719.1 | |
XM_017029231.1 | 225 | Intron | XP_016884720.1 |