Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGATCCCCAGCCAAAAATGGAC[C/T]ACGCTGGGAACGTGGCAGAGGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300688 | ||||||||||||||||||||
Literature Links: |
BCORL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCORL1 - BCL6 corepressor-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184772.2 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | NP_001171701.1 | |
NM_021946.4 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | NP_068765.3 | |
XM_005262452.4 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_005262509.2 | |
XM_005262453.4 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_005262510.2 | |
XM_005262454.3 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_005262511.2 | |
XM_005262455.4 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_005262512.2 | |
XM_005262456.4 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_005262513.2 | |
XM_006724776.3 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_006724839.1 | |
XM_006724777.3 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_006724840.1 | |
XM_006724779.2 | 846 | Missense Mutation | CAC,TAC | H,Y 3 | XP_006724842.1 | |
XM_017029721.1 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_016885210.1 | |
XM_017029722.1 | 846 | Missense Mutation | CAC,TAC | H,Y 101 | XP_016885211.1 |