Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTGGCCTTGTTTTGCAGGTGC[C/T]CTCCCGTTCTCTGCTGGCTCCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A43 - solute carrier family 25 member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145305.2 | 631 | Silent Mutation | GCC,GCT | A,A 174 | NP_660348.2 | |
XM_017029340.1 | 631 | Silent Mutation | GCC,GCT | A,A 174 | XP_016884829.1 | |
XM_017029341.1 | 631 | Silent Mutation | GCC,GCT | A,A 174 | XP_016884830.1 |