Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAACGTTCCCACTCACTCATGAG[C/G]TCAAAGTGTACAGGACGGCGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 301300 MIM: 300773 | ||||||||||||||||||||
Literature Links: |
ALAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALAS2 - 5'-aminolevulinate synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000032.4 | 1829 | Missense Mutation | GAC,GAG | D,E 565 | NP_000023.2 | |
NM_001037967.3 | 1829 | Missense Mutation | GAC,GAG | D,E 528 | NP_001033056.1 | |
NM_001037968.3 | 1829 | Missense Mutation | GAC,GAG | D,E 552 | NP_001033057.1 | |
XM_005261995.3 | 1829 | Missense Mutation | GAC,GAG | D,E 589 | XP_005262052.1 | |
XM_011530771.1 | 1829 | Missense Mutation | GAC,GAG | D,E 278 | XP_011529073.1 | |
XM_017029354.1 | 1829 | Intron | XP_016884843.1 |
APEX2 - apurinic/apyrimidinic endodeoxyribonuclease 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |