Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCACAAAGTCTTTGCTAGTCCA[A/G]TTTGGAGCTCTGAAGCAAAGATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEA11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEA11 - MAGE family member A11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011544.1 | Intron | NP_001011544.1 | ||||
NM_005366.4 | Intron | NP_005357.2 | ||||
XM_011531164.2 | Intron | XP_011529466.1 | ||||
XM_017029521.1 | Intron | XP_016885010.1 | ||||
XM_017029522.1 | Intron | XP_016885011.1 |