Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTAAATGGTAGATATGATCATG[C/T]CCCATTTGCTGATGAGGAAGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM120C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM120C - family with sequence similarity 120C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300788.1 | Intron | NP_001287717.1 | ||||
NM_017848.5 | Intron | NP_060318.4 | ||||
NM_198456.2 | Intron | NP_940858.2 | ||||
XM_006724589.3 | Intron | XP_006724652.1 |