Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAAACCACCAGGCTACCTCCTG[C/T]ACAACTGTCAATAAGACAACAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSL4 - acyl-CoA synthetase long-chain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318509.1 | 2530 | UTR 3 | NP_001305438.1 | |||
NM_001318510.1 | 2530 | UTR 3 | NP_001305439.1 | |||
NM_004458.2 | 2530 | UTR 3 | NP_004449.1 | |||
NM_022977.2 | 2530 | UTR 3 | NP_075266.1 | |||
XM_005262109.2 | 2530 | UTR 3 | XP_005262166.1 | |||
XM_006724635.2 | 2530 | UTR 3 | XP_006724698.1 | |||
XM_011530888.2 | 2530 | UTR 3 | XP_011529190.1 | |||
XM_011530889.2 | 2530 | UTR 3 | XP_011529191.1 |