Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGGCACTGGGGCCGGGGCCAC[C/T]GGGCTGGGCACAGGCGCAGCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC7 - armadillo repeat containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HN1 - hematological and neurological expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002032.2 | 682 | Missense Mutation | CAG,CGG | Q,R 126 | NP_001002032.1 | |
NM_001002033.2 | 682 | Silent Mutation | CCA,CCG | P,P 86 | NP_001002033.1 | |
NM_001288609.1 | 682 | Silent Mutation | CCA,CCG | P,P 86 | NP_001275538.1 | |
NM_001288610.1 | 682 | Silent Mutation | CCA,CCG | P,P 86 | NP_001275539.1 | |
NM_001288611.1 | 682 | UTR 3 | NP_001275540.1 | |||
NM_016185.3 | 682 | Silent Mutation | CCA,CCG | P,P 132 | NP_057269.1 |
NT5C - 5', 3'-nucleotidase, cytosolic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |