Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601117 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
THOP1 - thimet oligopeptidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF554 - zinc finger protein 554 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102651.1 | Intron | NP_001096121.1 | ||||
XM_006722635.3 | Intron | XP_006722698.1 | ||||
XM_011527660.2 | Intron | XP_011525962.1 | ||||
XM_011527662.2 | Intron | XP_011525964.1 | ||||
XM_011527664.2 | Intron | XP_011525966.1 | ||||
XM_011527667.2 | Intron | XP_011525969.1 | ||||
XM_017026234.1 | Intron | XP_016881723.1 | ||||
XM_017026235.1 | Intron | XP_016881724.1 | ||||
XM_017026236.1 | Intron | XP_016881725.1 |
Set Membership: |
HapMap |