Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602993 | ||||||||||||||||||||
Literature Links: |
LAIR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAIR2 - leukocyte associated immunoglobulin like receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002288.5 | 278 | Silent Mutation | CCA,CCG | P,P 52 | NP_002279.2 | |
NM_021270.4 | 278 | Silent Mutation | CCA,CCG | P,P 52 | NP_067154.1 | |
XM_011526961.2 | 278 | Silent Mutation | CCA,CCG | P,P 40 | XP_011525263.1 |