Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGGGAACCCTTGCCAAGAGCCG[C/T]GCTAAGTTAAATGGTAGAGCCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf45 - chromosome 11 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNJ5 - potassium voltage-gated channel subfamily J member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000890.3 | Intron | NP_000881.3 | ||||
XM_011542809.2 | Intron | XP_011541111.1 | ||||
XM_011542810.2 | Intron | XP_011541112.1 |