Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600859 MIM: 613635 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIMP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIMP2 - aminoacyl tRNA synthetase complex interacting multifunctional protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326606.1 | 4425 | Intron | NP_001313535.1 | |||
NM_001326607.1 | 4425 | Intron | NP_001313536.1 | |||
NM_001326609.1 | 4425 | Intron | NP_001313538.1 | |||
NM_001326611.1 | 4425 | Intron | NP_001313540.1 | |||
NM_006303.3 | 4425 | Intron | NP_006294.2 | |||
XM_005249847.3 | 4425 | Intron | XP_005249904.1 | |||
XM_005249848.2 | 4425 | Intron | XP_005249905.1 | |||
XM_005249850.3 | 4425 | Intron | XP_005249907.1 | |||
XM_006715772.2 | 4425 | Intron | XP_006715835.1 | |||
XM_011515519.1 | 4425 | Intron | XP_011513821.1 |
ANKRD61 - ankyrin repeat domain 61 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF2AK1 - eukaryotic translation initiation factor 2 alpha kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134335.1 | 4425 | UTR 3 | NP_001127807.1 | |||
NM_014413.3 | 4425 | UTR 3 | NP_055228.2 |