Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACCAGTGCAGAGATTAGTAAT[A/G]GCTGTTTTTACCCTAGAAGTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601443 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFL2 - cofilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243645.1 | 1505 | UTR 3 | NP_001230574.1 | |||
NM_021914.7 | 1505 | UTR 3 | NP_068733.1 | |||
NM_138638.4 | 1505 | UTR 3 | NP_619579.1 | |||
XM_011536363.2 | 1505 | UTR 3 | XP_011534665.1 |