Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACAGTTTGCTGTAAGATAACTT[C/T]CCGTGCATCTTTTAAATCAATGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613541 MIM: 602198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf65 - chromosome 12 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDK2AP1 - cyclin dependent kinase 2 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270433.1 | 1126 | Intron | NP_001257362.1 | |||
NM_001270434.1 | 1126 | Intron | NP_001257363.1 | |||
NM_004642.3 | 1126 | UTR 3 | NP_004633.1 |