Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTGTCATTAGGAATTCCTCCT[C/G]ATCAGCAGAGACTGATCTTTGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603766 MIM: 191343 | ||||||||||||||||||||
Literature Links: |
CLHC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLHC1 - clathrin heavy chain linker domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTIF2 - mitochondrial translational initiation factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS27A - ribosomal protein S27a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135592.2 | 338 | Missense Mutation | CAT,GAT | H,D 39 | NP_001129064.1 | |
NM_001177413.1 | 338 | Missense Mutation | CAT,GAT | H,D 39 | NP_001170884.1 | |
NM_002954.5 | 338 | Missense Mutation | CAT,GAT | H,D 39 | NP_002945.1 |