Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAAGGGAGTAGGAAAAACTTC[A/G]GAAATGTGTGGAATCTATAAGACTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614313 MIM: 611919 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACOT1 - acyl-CoA thioesterase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C14orf169 - chromosome 14 open reading frame 169 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HEATR4 - HEAT repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220484.1 | Intron | NP_001207413.1 | ||||
NM_203309.2 | Intron | NP_976054.2 | ||||
XM_006720143.2 | Intron | XP_006720206.1 | ||||
XM_011536760.2 | Intron | XP_011535062.1 | ||||
XM_011536761.2 | Intron | XP_011535063.1 | ||||
XM_017021289.1 | Intron | XP_016876778.1 |
Set Membership: |
HapMap |