Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGTGCCTACCTGCCAGTCTGTC[C/T]TGGGTGTGTCATTTGTCCATTTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605991 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf82 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2orf82 - chromosome 2 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206895.1 | Intron | NP_996778.1 | ||||
XM_017004080.1 | Intron | XP_016859569.1 | ||||
XM_017004081.1 | Intron | XP_016859570.1 | ||||
XM_017004082.1 | Intron | XP_016859571.1 | ||||
XM_017004083.1 | Intron | XP_016859572.1 | ||||
XM_017004084.1 | Intron | XP_016859573.1 |
NGEF - neuronal guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |