Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182870 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLEKHG3 - pleckstrin homology and RhoGEF domain containing G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308147.1 | 8114 | Intron | NP_001295076.1 |
SPTB - spectrin beta, erythrocytic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000347.5 | 8114 | Intron | NP_000338.3 | |||
NM_001024858.2 | 8114 | UTR 3 | NP_001020029.1 | |||
XM_005268023.4 | 8114 | UTR 3 | XP_005268080.1 | |||
XM_011537105.2 | 8114 | Intron | XP_011535407.1 | |||
XM_017021612.1 | 8114 | UTR 3 | XP_016877101.1 | |||
XM_017021613.1 | 8114 | Intron | XP_016877102.1 | |||
XM_017021614.1 | 8114 | Intron | XP_016877103.1 |
Set Membership: |
HapMap |