Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATTATAAGGACTGTGGCATAAA[T/C]TTTTAAATGAGTTATATTGAAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603623 | ||||||||||||||||||||
Literature Links: |
KIAA1522 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1522 - KIAA1522 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198972.1 | 1671 | Intron | NP_001185901.1 | |||
NM_001198973.1 | 1671 | Intron | NP_001185902.1 | |||
NM_020888.2 | 1671 | Intron | NP_065939.2 | |||
XM_006710791.3 | 1671 | Intron | XP_006710854.1 | |||
XM_017001916.1 | 1671 | Intron | XP_016857405.1 | |||
XM_017001917.1 | 1671 | Intron | XP_016857406.1 |
YARS - tyrosyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003680.3 | 1671 | UTR 3 | NP_003671.1 | |||
XM_011542347.1 | 1671 | UTR 3 | XP_011540649.1 | |||
XM_011542348.1 | 1671 | UTR 3 | XP_011540650.1 | |||
XM_017002651.1 | 1671 | UTR 3 | XP_016858140.1 |