Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGGGGATACCCAAATGAAGAAT[A/T]TGAAGTTGTGACTGAAGATGGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPF - lipase F, gastric type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198828.1 | 202 | Missense Mutation | TAT,TTT | Y,F 49 | NP_001185757.1 | |
NM_001198829.1 | 202 | Missense Mutation | TAT,TTT | Y,F 59 | NP_001185758.1 | |
NM_001198830.1 | 202 | Missense Mutation | TAT,TTT | Y,F 59 | NP_001185759.1 | |
NM_004190.3 | 202 | Missense Mutation | TAT,TTT | Y,F 49 | NP_004181.1 | |
XM_011540311.1 | 202 | Missense Mutation | TAT,TTT | Y,F 49 | XP_011538613.1 |