Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCTAGCACCCCCTCTGCAGGTG[C/T]TGCACCCCTCATCCTGTCTCGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK3 - kallikrein related peptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030047.1 | 91 | Missense Mutation | GCT,GTT | A,V 17 | NP_001025218.1 | |
NM_001030048.1 | 91 | Missense Mutation | GCT,GTT | A,V 17 | NP_001025219.1 | |
NM_001648.2 | 91 | Missense Mutation | GCT,GTT | A,V 17 | NP_001639.1 |
LOC105372441 - uncharacterized LOC105372441 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |