Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGGCACCGCCCAGGGCGCCCC[C/T]AGCGGAGGCGGCGGCTGCAAGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM67 - tripartite motif containing 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004342.3 | 681 | Silent Mutation | CCC,CCT | P,P 275 | NP_001004342.3 | |
NM_001300889.1 | 681 | Silent Mutation | CCC,CCT | P,P 213 | NP_001287818.1 | |
XM_011544192.2 | 681 | Silent Mutation | CCC,CCT | P,P 275 | XP_011542494.1 | |
XM_017001323.1 | 681 | Silent Mutation | CCC,CCT | P,P 275 | XP_016856812.1 |