Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCGCACCGTTTGGCTCCTGCTG[C/G]ACGAATGGGTAGAAGCTATTGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609076 MIM: 607882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL6 - F-box and leucine rich repeat protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012162.3 | 299 | Intron | NP_036294.2 | |||
NM_024555.5 | 299 | Intron | NP_078831.4 |
SLC52A2 - solute carrier family 52 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM249 - transmembrane protein 249 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252402.2 | 299 | Silent Mutation | GTC,GTG | V,V 45 | NP_001239331.1 | |
NM_001252404.2 | 299 | Silent Mutation | GTC,GTG | V,V 45 | NP_001239333.1 | |
NM_001280561.1 | 299 | Silent Mutation | GTC,GTG | V,V 45 | NP_001267490.1 |