Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATGGTAGGAGTGCAGCTTTCT[C/T]CCCCAGTTACTTTCCAGCTCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603725 MIM: 608073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF17 - fibroblast growth factor 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPM2 - nucleophosmin/nucleoplasmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286680.1 | 952 | Missense Mutation | CCC,TCC | P,S 100 | NP_001273609.1 | |
NM_001286681.1 | 952 | Missense Mutation | CCC,TCC | P,S 100 | NP_001273610.1 | |
NM_182795.1 | 952 | Missense Mutation | CCC,TCC | P,S 100 | NP_877724.1 | |
XM_011544360.2 | 952 | Missense Mutation | CCC,TCC | P,S 175 | XP_011542662.1 | |
XM_011544362.2 | 952 | Missense Mutation | CCC,TCC | P,S 100 | XP_011542664.1 | |
XM_011544363.2 | 952 | Missense Mutation | CCC,TCC | P,S 100 | XP_011542665.1 | |
XM_011544364.2 | 952 | Missense Mutation | CCC,TCC | P,S 42 | XP_011542666.1 | |
XM_017012947.1 | 952 | Missense Mutation | CCC,TCC | P,S 133 | XP_016868436.1 | |
XM_017012948.1 | 952 | Missense Mutation | CCC,TCC | P,S 100 | XP_016868437.1 | |
XM_017012949.1 | 952 | Missense Mutation | CCC,TCC | P,S 100 | XP_016868438.1 | |
XM_017012950.1 | 952 | Missense Mutation | CCC,TCC | P,S 58 | XP_016868439.1 |