Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAATCTTACCTGAATCCAGTCTC[C/T]GAGGTAATATTGCTGGGGTAAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM192 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM192 - transmembrane protein 192 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100389.1 | 803 | Silent Mutation | TCA,TCG | S,S 221 | NP_001093859.1 | |
XM_011531717.2 | 803 | Silent Mutation | TCA,TCG | S,S 221 | XP_011530019.1 | |
XM_011531718.2 | 803 | Silent Mutation | TCA,TCG | S,S 176 | XP_011530020.1 |