Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATTTTGTGTCTTTTCTGGACTC[A/G]GATGACCCAGAGTCCCATGGCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609738 MIM: 608589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF9 - immunoglobulin superfamily member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01133 - long intergenic non-protein coding RNA 1133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLAMF9 - SLAM family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146172.1 | 616 | Nonsense Mutation | CGA,TGA | R,* 167 | NP_001139644.1 | |
NM_001146173.1 | 616 | UTR 3 | NP_001139645.1 | |||
NM_033438.3 | 616 | Nonsense Mutation | CGA,TGA | R,* 258 | NP_254273.2 | |
XM_017002756.1 | 616 | Nonsense Mutation | CGA,TGA | R,* 288 | XP_016858245.1 |