Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACGCTATATCTGTTTTGAGTTAT[A/G]TATCTGCAGCAGCTGGTTAAAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADH7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADH7 - alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000673.4 | 1726 | UTR 3 | NP_000664.2 | |||
NM_001166504.1 | 1726 | UTR 3 | NP_001159976.1 |