Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGTCCAGAGATAAATGAAGGCTG[C/T]TGTTAACAAGCATGACAGAAAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2L10 - BCL2 like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306168.1 | 629 | Missense Mutation | AAC,AGC | N,S 217 | NP_001293097.1 | |
NM_020396.3 | 629 | Missense Mutation | ACA,GCA | T,A 194 | NP_065129.1 |