Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGGCTTATGTAGGCGTAGAAGA[C/T]AGCGGTGTAATACATGCACACCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR13A1 - olfactory receptor family 13 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004297.2 | 4899 | Missense Mutation | ATC,GTC | I,V 273 | NP_001004297.2 | |
XM_011540147.2 | 4899 | Missense Mutation | ATC,GTC | I,V 273 | XP_011538449.1 | |
XM_017016632.1 | 4899 | Missense Mutation | ATC,GTC | I,V 273 | XP_016872121.1 | |
XM_017016633.1 | 4899 | Missense Mutation | ATC,GTC | I,V 273 | XP_016872122.1 | |
XM_017016634.1 | 4899 | Missense Mutation | ATC,GTC | I,V 273 | XP_016872123.1 |