Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGCCAGAACCTTCTTTCTCAC[A/G]GGAAGACTGTGGTACCAGTGCGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610602 MIM: 191525 MIM: 612492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH2 - alkB homolog 2, alpha-ketoglutarate dependent dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001655.2 | 1110 | Silent Mutation | CCC,CCT | P,P 239 | NP_001001655.1 | |
NM_001145374.1 | 1110 | Silent Mutation | CCC,CCT | P,P 239 | NP_001138846.1 | |
NM_001145375.1 | 1110 | Silent Mutation | CCC,CCT | P,P 239 | NP_001138847.1 | |
NM_001205179.1 | 1110 | UTR 3 | NP_001192108.1 | |||
NM_001205180.1 | 1110 | UTR 3 | NP_001192109.1 | |||
XM_005253835.4 | 1110 | Silent Mutation | CCC,CCT | P,P 239 | XP_005253892.1 | |
XM_005253836.1 | 1110 | UTR 3 | XP_005253893.1 |
UNG - uracil DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP30 - ubiquitin specific peptidase 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301175.1 | 1110 | Intron | NP_001288104.1 | |||
NM_032663.4 | 1110 | Intron | NP_116052.2 | |||
XM_005253962.3 | 1110 | Intron | XP_005254019.1 | |||
XM_005253965.4 | 1110 | Intron | XP_005254022.1 | |||
XM_006719653.3 | 1110 | Intron | XP_006719716.1 | |||
XM_011538894.2 | 1110 | Intron | XP_011537196.1 | |||
XM_017020048.1 | 1110 | Intron | XP_016875537.1 | |||
XM_017020049.1 | 1110 | Intron | XP_016875538.1 | |||
XM_017020050.1 | 1110 | Intron | XP_016875539.1 | |||
XM_017020051.1 | 1110 | Intron | XP_016875540.1 | |||
XM_017020052.1 | 1110 | Intron | XP_016875541.1 | |||
XM_017020053.1 | 1110 | Intron | XP_016875542.1 | |||
XM_017020054.1 | 1110 | Intron | XP_016875543.1 |