Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGACATGGCGGTTACCCTGAGTC[C/T]CTTGCTGGGCGGGCGCGTTTGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602293 MIM: 616718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIB1 - calcium and integrin binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277764.1 | Intron | NP_001264693.1 | ||||
NM_006384.3 | Intron | NP_006375.2 | ||||
XM_006720375.1 | Intron | XP_006720438.1 |
LOC400446 - PERP, TP53 apoptosis effector pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NGRN - neugrin, neurite outgrowth associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033088.1 | Intron | NP_001028260.2 |
TTLL13P - tubulin tyrosine ligase like 13, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |