Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTCAGCGCCCTAGGTCATGAG[A/G]ATTTCGATGTCAAGCTCCAGCCGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
IQCF6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IQCF6 - IQ motif containing F6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143833.3 | 372 | Silent Mutation | ATC,ATT | I,I 104 | NP_001137305.2 | |
XM_005265168.3 | 372 | Silent Mutation | ATC,ATT | I,I 124 | XP_005265225.1 | |
XM_011533728.1 | 372 | Silent Mutation | ATC,ATT | I,I 104 | XP_011532030.1 |