Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCGACAGCTGCGTGTCTCCCA[C/T]GGCTGCGTCTCCAAGATTCTTTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAX7 - paired box 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135254.1 | 838 | Silent Mutation | CAC,CAT | H,H 80 | NP_001128726.1 | |
NM_002584.2 | 838 | Silent Mutation | CAC,CAT | H,H 80 | NP_002575.1 | |
NM_013945.2 | 838 | Silent Mutation | CAC,CAT | H,H 80 | NP_039236.1 |