Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGAGTAGTGCAAGCCTATAGC[A/G]TCCTCTGTCCTCCCCTCTCCTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLFN14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLFN14 - schlafen family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129820.1 | 2599 | Missense Mutation | CGC,TGC | R,C 827 | NP_001123292.1 | |
XM_017024576.1 | 2599 | Missense Mutation | CGC,TGC | R,C 827 | XP_016880065.1 | |
XM_017024577.1 | 2599 | Missense Mutation | CGC,TGC | R,C 827 | XP_016880066.1 | |
XM_017024578.1 | 2599 | Missense Mutation | CGC,TGC | R,C 827 | XP_016880067.1 | |
XM_017024579.1 | 2599 | Missense Mutation | CGC,TGC | R,C 827 | XP_016880068.1 |