Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTTGGGCGTGCCCTACGCCAC[A/G]CCGCCCCTGGGCGCCCGCCGCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLGN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLGN2 - neuroligin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020795.3 | 301 | Silent Mutation | ACA,ACG | T,T 76 | NP_065846.1 | |
XM_005256744.3 | 301 | Silent Mutation | ACA,ACG | T,T 76 | XP_005256801.1 | |
XM_017024897.1 | 301 | Intron | XP_016880386.1 |
TMEM256 - transmembrane protein 256 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM256-PLSCR3 - TMEM256-PLSCR3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |