Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAATTATAACCCACACATCTTC[C/T]TCTGCATTTGTGACTTCATTCACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611676 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCL2 - phosducin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152401.2 | 427 | Silent Mutation | GAA,GAG | E,E 108 | NP_689614.2 | |
XM_005265728.3 | 427 | Silent Mutation | GAA,GAG | E,E 108 | XP_005265785.1 | |
XM_011534374.2 | 427 | Silent Mutation | GAA,GAG | E,E 76 | XP_011532676.1 | |
XM_017007745.1 | 427 | Silent Mutation | GAA,GAG | E,E 108 | XP_016863234.1 |