Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTGCTTAAACACAAGGCCAGG[A/G]TGGATGTTGCTGATAAGGTAAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKDD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKDD1B - ankyrin repeat and death domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276713.1 | 725 | Missense Mutation | ATG,GTG | M,V 127 | NP_001263642.1 | |
XM_011543618.2 | 725 | Missense Mutation | ATG,GTG | M,V 127 | XP_011541920.1 | |
XM_011543623.2 | 725 | Intron | XP_011541925.1 | |||
XM_017009814.1 | 725 | Missense Mutation | ATG,GTG | M,V 127 | XP_016865303.1 | |
XM_017009815.1 | 725 | Missense Mutation | ATG,GTG | M,V 127 | XP_016865304.1 | |
XM_017009816.1 | 725 | Missense Mutation | ATG,GTG | M,V 127 | XP_016865305.1 | |
XM_017009817.1 | 725 | Missense Mutation | ATG,GTG | M,V 127 | XP_016865306.1 |