Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGCTCTGTGGCGTTTCATCCC[A/G]CTCTCCCAGGCACAAGACACAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929715 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929715 - uncharacterized LOC101929715 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODC1 - ornithine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287188.1 | 1896 | Silent Mutation | AGC,AGT | S,S 316 | NP_001274117.1 | |
NM_001287189.1 | 1896 | Silent Mutation | AGC,AGT | S,S 445 | NP_001274118.1 | |
NM_001287190.1 | 1896 | Silent Mutation | AGC,AGT | S,S 445 | NP_001274119.1 | |
NM_002539.2 | 1896 | Silent Mutation | AGC,AGT | S,S 445 | NP_002530.1 |
SNORA80B - small nucleolar RNA, H/ACA box 80B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |