Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCGAACGGCTAAAGGCCGGGGG[A/T]CCGCGCTTAAGGCCCCCGGCACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142967 MIM: 142966 | ||||||||||||||||||||
Literature Links: |
HOXB-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXB-AS1 - HOXB cluster antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB2 - homeobox B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002145.3 | 896 | Missense Mutation | GAC,GTC | D,V 259 | NP_002136.1 | |
XM_005257275.4 | 896 | Missense Mutation | GAC,GTC | D,V 150 | XP_005257332.1 |
HOXB3 - homeobox B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |