Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAGTCTGTTGCTGTGATTGTG[C/T]TGGACTTGCTGCTTGTGTCTGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VN1R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VN1R1 - vomeronasal 1 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020633.3 | 1015 | Silent Mutation | CAA,CAG | Q,Q 254 | NP_065684.1 |
ZNF749 - zinc finger protein 749 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |