Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCGCTGAGCCCGCCTTTGCTGG[C/T]TGCTGGTGGAGACGGTGTCTACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCN1 - lipocalin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252617.1 | 1180 | Intron | NP_001239546.1 | |||
NM_001252618.1 | 1180 | Intron | NP_001239547.1 | |||
NM_001252619.1 | 1180 | Intron | NP_001239548.1 | |||
NM_002297.3 | 1180 | Intron | NP_002288.1 | |||
XM_011518675.2 | 1180 | UTR 3 | XP_011516977.1 |